UV detector, 280
Lieferant: Spectrum
SPEC142602EA 5580 EUR
SPEC142602
UV detector, 280
UV detector with fixed wavelength flow-through spectrophotometer for use as a chromatographic detector. It works with HPLC and low pressure systems and features a direct reading digital output.
To determine the concentrations of compounds as they elute from the column and provides information on the condition of the column and on the identity of the sample components.
Lieferumfang: Includes a pathlength flow cell (5 mm), filters and tubing fittings.