1-Vinyl-2-pyrrolidon
Lieferant: LGC Standards PROMOCHEM
PROOMM0083.03EA
1230
EUR
PROOMM0083.03
1-Vinyl-2-pyrrolidon
1-Vinyl-2-pyrrolidon
Formel:
C₆H₉NO Molekulargewicht: 111,1418 g/mol |
MDL Number:
MFCD00003197 CAS-Nummer: 88-12-0 EINECS: 201-800-4 UN-Nummer: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...