Inoculating loops with needle end, Sterilin®
Lieferant: Thermo Scientific
391-0447EA 140 EUR
SLINSL1H
Inoculating loops with needle end, Sterilin®
Plastic loops with needle ends for dilution streaking and obtaining isolated colonies. Available with a choice of rigid or flexible loops to suit different applications and user preferences.
  • Features a fixed volume loop at one end and a needle for colony extraction at the other
  • Accuracy of ±20% certified using Evans Blue Dye Method (calibration certificate provided)
  • No rough edges on the loop head means smooth problem free plating and streaking of cultures
  • Free of lubricants/oils and electrostatic charges to facilitate consistent wetting and complete liquid transfer
  • Hexagonal shaft improves grip and assists orientation