(Ethoxycarbonylmethyl)triphenylphosphoniumbromid

Lieferant: Molekula

MOLEM87142634EA 0 EUR
MOLEM87142634
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
Formel: C₂₂H₂₂BrO₂P
Molekulargewicht: 429,30 g/mol
MDL Number: MFCD00011835
CAS-Nummer: 1530-45-6
EINECS: 216-230-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR