(R)-(+)-Tetrahydrofuran-2-carbonsäure

Lieferant: Molekula

MOLE40022020-1GEA 0 EUR
MOLE40022020-1G
(R)-(+)-Tetrahydrofuran-2-carbonsäure
(R)-(+)-Tetrahydrofuran-2-carbonsäure
Formel: C₅H₈O₃
Molekulargewicht: 116,12 g/mol
MDL Number: MFCD00211271
CAS-Nummer: 87392-05-0
UN-Nummer: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR