(4-Nitrophenyl)-α-D-galactopyranosid

Lieferant: Molekula

MOLEM42887890EA 0 EUR
MOLEM42887890
(4-Nitrophenyl)-α-D-galactopyranosid
(4-Nitrophenyl)-α-D-galactopyranosid
Formel: C₁₂H₁₅NO₈
Molekulargewicht: 301,26 g/mol
MDL Number: MFCD00065050
CAS-Nummer: 7493-95-0
EINECS: 231-343-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR