N-Methylbistrifluoracetamid

Gefahr

HYPETS-49703EA 364 EUR
HYPETS-49703 HYPETS-49701
N-Methylbistrifluoracetamid
N-Methylbistrifluoracetamid
Formel: C₅H₃F₆NO₂
Molekulargewicht: 223,07 g/mol
MDL Number: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR