2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Lieferant: Molekula
MOLEM76880547EA
0
EUR
MOLEM76880547
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel:
C₂H₅NaO₃S₂ Molekulargewicht: 164,18 g/mol |
MDL Number:
MFCD00007535 CAS-Nummer: 19767-45-4 EINECS: 243-285-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...