Decyl-β-D-thioglucopyranosid

Lieferant: Molekula

MOLE35887761-250MGEA 0 EUR
MOLE35887761-250MG
Decyl-β-D-thioglucopyranosid
Decyl-β-D-thioglucopyranosid
Formel: C₁₆H₃₂O₅S
CAS-Nummer: 98854-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR