Ethoxyquin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

31519-250MGEA 62.7 EUR
31519-250MG
Ethoxyquin, Supelco®
Ethoxyquin
Formel: C₁₄H₁₉NO
MDL Number: MFCD00023883
CAS-Nummer: 91-53-2
EINECS: 202-075-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR