Allyltriphenylphosphoniumchlorid

Lieferant: Molekula

MOLE31295750-25GEA 0 EUR
MOLE31295750-25G
Allyltriphenylphosphoniumchlorid
Allyltriphenylphosphoniumchlorid
Formel: C₂₁H₂₀ClP
Molekulargewicht: 338,82 g/mol
MDL Number: MFCD00031542
CAS-Nummer: 18480-23-4
EINECS: 242-368-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR