Tetraethylorthocarbonat

Lieferant: Molekula

MOLEM81219845EA 0 EUR
MOLEM81219845
Tetraethylorthocarbonat
Tetraethylorthocarbonat
Formel: C₉H₂₀O₄
Molekulargewicht: 192,26 g/mol
MDL Number: MFCD00009221
CAS-Nummer: 78-09-1
EINECS: 201-082-2
UN-Nummer: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR