Phenyltrimethylphosphoniumiodid

Lieferant: Thermo Scientific

MAYBCD00125.1EA 0 EUR
MAYBCD00125.1 CHMPCD00125.250
Phenyltrimethylphosphoniumiodid
Phenyltrimethylphosphonium iodide
Formel: C₉H₁₄IP
Molekulargewicht: 280,089 g/mol
MDL Number: MFCD00050202
CAS-Nummer: 1006-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR