2-Pyridylacetonitril

Lieferant: Molekula

MOLEM57322098EA 0 EUR
MOLEM57322098
2-Pyridylacetonitril
2-Pyridylacetonitril
Formel: C₇H₆N₂
Molekulargewicht: 118,14 g/mol
MDL Number: MFCD00006346
CAS-Nummer: 2739-97-1
EINECS: 220-364-6
UN-Nummer: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR