N,N-Dimethylcarbamoylchlorid

Lieferant: Molekula

MOLEM64798950EA 0 EUR
MOLEM64798950
N,N-Dimethylcarbamoylchlorid
N,N-Dimethylcarbamoylchlorid
Formel: C₃H₆ClNO
Molekulargewicht: 107,54 g/mol
MDL Number: MFCD00000635
CAS-Nummer: 79-44-7
EINECS: 201-208-6
UN-Nummer: 2262
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR