L(-)-Methioninol

Lieferant: Molekula

MOLE64837537-1GEA 0 EUR
MOLE64837537-1G
L(-)-Methioninol
L(-)-Methioninol
Formel: C₅H₁₃NOS
Molekulargewicht: 135,2 g/mol
CAS-Nummer: 2899-37-8
EINECS: 220-788-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR