1-Acetyl-2-phenylhydrazin

Lieferant: Molekula

MOLEM69389939EA 0 EUR
MOLEM69389939
1-Acetyl-2-phenylhydrazin
1-Acetyl-2-phenylhydrazin
Formel: C₈H₁₀N₂O
Molekulargewicht: 150,18 g/mol
MDL Number: MFCD00008672
CAS-Nummer: 114-83-0
EINECS: 204-055-3
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR