4,4'-Dimethoxytritylchlorid

Lieferant: Molekula

MOLEM10955409EA 0 EUR
MOLEM10955409
4,4'-Dimethoxytritylchlorid
4,4'-Dimethoxytritylchlorid
Formel: C₂₁H₁₉ClO₂
Molekulargewicht: 338,8 g/mol
MDL Number: MFCD00008409
CAS-Nummer: 40615-36-9
EINECS: 255-002-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR