8-Chinolinsulfonylchlorid

Lieferant: Molekula

MOLEM74012020EA 0 EUR
MOLEM74012020
8-Chinolinsulfonylchlorid
8-Chinolinsulfonylchlorid
Formel: C₉H₆ClNO₂S
Molekulargewicht: 227,67 g/mol
MDL Number: MFCD00006808
CAS-Nummer: 18704-37-5
EINECS: 242-515-5
UN-Nummer: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR