Dibenzyldiisopropylphosphoramidit

Lieferant: Molekula

MOLE17862617-5GEA 88.3 EUR
MOLE17862617-5G
Dibenzyldiisopropylphosphoramidit
Dibenzyldiisopropylphosphoramidit
Formel: C₂₀H₂₈NO₂P
Molekulargewicht: 345,43
MDL Number: MFCD00191988
CAS-Nummer: 108549-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR