Phthalazin

Lieferant: Molekula

MOLE70475097-5GEA 88.3 EUR
MOLE70475097-5G
Phthalazin
Phthalazine
Formel: C₈H₆N₂
Molekulargewicht: 130,15
MDL Number: MFCD00006908
CAS-Nummer: 253-52-1
EINECS: 205-963-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR