Benzyltrimethylammoniumdichloroiodat

Lieferant: Molekula

MOLE85078328-5GEA 0 EUR
MOLE85078328-5G
Benzyltrimethylammoniumdichloroiodat
Benzyltrimethylammoniumdichloroiodat
Formel: C₁₀H₁₆Cl₂IN
Molekulargewicht: 348,06 g/mol
MDL Number: MFCD00075259
CAS-Nummer: 114971-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR