Tetrabutylammoniumdihydrogenphosphat ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
19261.04EA
105
EUR
19261.04
19261.02
19261.09
19261.18
Tetrabutylammoniumdihydrogenphosphat ≥99%
Tetrabutylammoniumdihydrogenphosphat
Used as a counter-ion in ion-pair chromatography of water-soluble vitamins
Formel:
C₁₆H₃₈NO₄P Molekulargewicht: 339,46 g/mol Schmelzpunkt: 151…154 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064526 CAS-Nummer: 5574-97-0 EINECS: 226-947-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...