5-Methyl-2-pyrazincarbonsäure

Lieferant: Molekula

MOLEM75876957EA 77.4 EUR
MOLEM75876957
5-Methyl-2-pyrazincarbonsäure
5-Methylpyrazine-2-carboxylic acid
Formel: C₆H₆N₂O₂
Molekulargewicht: 138,13
MDL Number: MFCD00068241
CAS-Nummer: 5521-55-1
EINECS: 413-260-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR