N-Acetyl-L-methionin

Lieferant: Molekula

MOLE22003320-100GEA 151 EUR
MOLE22003320-100G
N-Acetyl-L-methionin
N-Acetyl-L-methionin
Formel: C₇H₁₃NO₃S
Molekulargewicht: 191,2 g/mol
CAS-Nummer: 65-82-7
EINECS: 200-617-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR