Bithionol, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

31622-250MGEA 53.6 EUR
31622-250MG
Bithionol, Supelco®
Bithionol
Formel: C₁₂H₆Cl₄O₂S
Molekulargewicht: 356,0518 g/mol
MDL Number: MFCD00055727
CAS-Nummer: 97-18-7
EINECS: 202-565-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR