Thiophen-3-acetonitril

Lieferant: Molekula

MOLE46123285-25GEA 0 EUR
MOLE46123285-25G
Thiophen-3-acetonitril
Thiophen-3-acetonitril
Formel: C₆H₅NS
Molekulargewicht: 123,18 g/mol
MDL Number: MFCD00005471
CAS-Nummer: 13781-53-8
EINECS: 237-433-1
UN-Nummer: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR