(±)-1,1′-Bi-2-naphthol

Lieferant: Molekula

MOLEM22387835EA 0 EUR
MOLEM22387835
(±)-1,1′-Bi-2-naphthol
(±)-1,1'-Bi-2-naphthol
Formel: C₂₀H₁₄O₂
Molekulargewicht: 286,33 g/mol
MDL Number: MFCD00004068
CAS-Nummer: 602-09-5
EINECS: 210-014-0
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR