Label printer systems, benchtop, IP™
Lieferant: Brady
BRDY360815EA 1645 EUR
BRDY360815 BRDY360817 BRDY360844
Label printer systems, benchtop, IP™
Brady IP™ printer is a fully integrated, easy-to-use printer, material, and software system. Smart Cell technology allows the printer, materials, and ribbon to communicate with each other and also it provides material recognition, automatic formatting, and user-friendly printer features for labels, sleeves, and tags.
  • USB, Serial and optional WiFi, Ethernet connectivity
  • Easy material loading
  • Intuitive with label design software
  • Detects loaded material and ribbon and displays the user information on the LCD
  • Used for asset identification, barcode labelling, wire and cable marking and circuit board identification
  • 20 cm/second print speed
Lieferumfang: Printer is supplied with a USB cable, drivers CD, power cord and a codesoft basic CD.