3-Amino-2-methylbenzotrifluorid

Lieferant: Molekula

MOLEM54855298EA 0 EUR
MOLEM54855298
3-Amino-2-methylbenzotrifluorid
3-Amino-2-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molekulargewicht: 175,16 g/mol
MDL Number: MFCD00153216
CAS-Nummer: 54396-44-0
EINECS: 259-145-5
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR