5-Azacytosin

Lieferant: Molekula

MOLE35283847-5GEA 0 EUR
MOLE35283847-5G
5-Azacytosin
5-Azacytosin
Formel: C₃H₄N₄O
Molekulargewicht: 112,09 g/mol
MDL Number: MFCD00149402
CAS-Nummer: 931-86-2
EINECS: 213-242-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR