N-Acetyl-D-leucin

Lieferant: Molekula

MOLE35738612-1GEA 0 EUR
MOLE35738612-1G
N-Acetyl-D-leucin
N-Acetyl-D-leucin
Formel: C₈H₁₅NO₃
Molekulargewicht: 173,2 g/mol
MDL Number: MFCD00066069
CAS-Nummer: 19764-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR