(±)-α-Methyltryptamin

Lieferant: Molekula

MOLEM70475563EA 0 EUR
MOLEM70475563
(±)-α-Methyltryptamin
(±)-α-Methyltryptamin
Formel: C₁₁H₁₄N₂
CAS-Nummer: 299-26-3
EINECS: 206-073-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR