2-(4-Chlorphenoxy)isobuttersäure

Lieferant: Molekula

MOLE69210117-100GEA 0 EUR
MOLE69210117-100G
2-(4-Chlorphenoxy)isobuttersäure
2-(4-Chlorphenoxy)isobuttersäure
Formel: C₁₀H₁₁ClO₃
Molekulargewicht: 214,65 g/mol
MDL Number: MFCD00004192
CAS-Nummer: 882-09-7
EINECS: 212-925-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR