3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäure

Lieferant: Molekula

MOLEM70030628EA 0 EUR
MOLEM70030628
3-(2-Chlorphenyl)-5-methyl-4-isoxazolcarbonsäure
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₈ClNO₃
Molekulargewicht: 237,64 g/mol
MDL Number: MFCD00020813
CAS-Nummer: 23598-72-3
EINECS: 245-770-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR