Ethyl-2-oxocyclopentancarboxylat

Lieferant: Molekula

MOLE14292361-25GEA 69.5 EUR
MOLE14292361-25G
Ethyl-2-oxocyclopentancarboxylat
Ethyl-2-oxocyclopentancarboxylat
Formel: C₈H₁₂O₃
Molekulargewicht: 156,18
MDL Number: MFCD00001412
CAS-Nummer: 611-10-9
EINECS: 210-253-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR